Viton® O-Rings, Chemglass
# de catalogue 89029-269
Fournisseur: Chemglass
Small Business Enterprise
Spécifications
- Cat. no.:89029-269
- Supplier No.:CG-305-243
Spécifications
A propos de cet article
Brown, low compression, 75 durometer Viton® O-rings are suitable for acids, aliphatic hydrocarbons, oxidizing agents, and all petroleum products
Note: Viton® O-ring 89029-223 is not a standard size.
They are especially good for vacuum service and low gas permeability. Temperature range: –30 to 200°C (–22 to 392°F).
Customers Who Bought This Also Bought