O-Rings, Perfluoro, Chemglass
# de catalogue 89027-254
Fournisseur: Chemglass
Small Business Enterprise
Spécifications
- Description:Perfluoro O-rings, white
- Cat. no.:89027-254
- Supplier No.:CG-309-009
Spécifications
A propos de cet article
Perfluoro is a family of perfluoroelastomer compounds designed to provide superior chemical resistance, similar O-rings such as Kalrez® 4079 and Chemraz®.
- Perfluoro O-rings are stocked in WHITE 75 durometer
- Effective service temperature is –7 to 230 °C
- Used with Duran flanges
These O-rings are used with acids, inorganic bases, alcohols/ethers, aromatic hydro-carbons, esters, aldehydes/ ketones, aliphatic hydrocarbons, amines and chlorocarbon solvents.
Customers Who Bought This Also Bought