ContactUsLinkComponent
VWR* PTFE Solid Stopper
VWR® Flathead [ST] Stoppers,PTFE
Catalog # 89026-340
Supplier:  VWR International
VWR* PTFE Solid Stopper
VWR® Flathead [ST] Stoppers,PTFE
Catalog # 89026-340
Supplier:  VWR International
Specifications
  • ST Joint No.:
    24/40
  • Height:
    55 mm
  • Cat. no.:
    89026-340
Specifications
About this item
Inert and chemically resistant solid PTFE stoppers for use with standard ground glass joints.
  • Chemically resistant
  • Inert
  • PTFE fabrication
Ideal for sealing glass containers and glass jointware assemblies. Trapezoidal sealing rings provide high vacuum performance and reduce seizure, while the non-stick properties ensure easy removal. Usable to 280°C (536°F).
Customers Who Bought This Also Bought