VWR® Magnetic Stir Bar Retrievers, PTFE
# de catalogue 89026-260
Fournisseur: VWR International
Spécifications
- Coating:PTFE
- Ø:10 mm (3/8")
- Length:250 mm (927/32")
- Description:Retrievers with Alnico V Magnet
- Cat. no.:89026-260
Spécifications
A propos de cet article
Molded in pure, FDA-approved PTFE, these retrievers are inert and non-contaminating.
- Inert
- PTFE fabrication
- Autoclavable
- Magnetic cores available in different variants
Available with standard Alnico V or samarium cobalt rare earth magnetic core. Autoclavable.
Customers Who Bought This Also Bought