Order Entry
ContactUsLinkComponent
Hirschmann* volumetric glass flask
Volumetric Flasks with [ST] PTFE Stopper, Class A, Hirschmann
# de catalogue 89025-830
Fournisseur:  Hirschmann
Hirschmann* volumetric glass flask
Volumetric Flasks with [ST] PTFE Stopper, Class A, Hirschmann
# de catalogue 89025-830
Fournisseur:  Hirschmann

Spécifications

  • Capacity:
    250 ml
  • Cat. no.:
    89025-830
  • ST Stopper No.:
    16
  • Tolerance:
    ±0.12 ml
  • Supplier No.:
    282-T-250

Spécifications

A propos de cet article

Flasks feature a fine white graduation line and marking spot

Meet ASTM E288, Class A requirements.

Graduated to contain. Flasks include a [ST] PTFE stopper.

Customers Who Bought This Also Bought