ContactUsLinkComponent
Waste Gas Control Supplies, VetEquip®
Catalogue # 89012-608
Fournisseur:  Vetequip
undefined
Waste Gas Control Supplies, VetEquip®
Catalogue # 89012-608
Fournisseur:  Vetequip
Supplier Number:  931401
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Spécifications
  • Description:
    Activated charcoal filter VaporGuard
  • Cat. no.:
    89012-608
  • Supplier No.:
    931401
Spécifications
À propos de cet article
The scavenging interface kit features an interface valve that is installed between an anesthesia machine and a vacuum evacuation system. The kit helps control and monitor exposure to anesthetic waste gases.
  • Valve equalizes both positive and negative pressure so that no metabolic gases are pulled past the patient
  • Monitoring badge adsorbs halogenated hydrocarbons, including isoflurane and halothene
  • After <=8 hours of exposure, the badge should be sent to a chemical laboratory for analysis of exposure levels
The activated charcoal adsorption filter adsorbs passively and has been proven effective in eliminating waste anesthetic gas pollution.
Ordering information: Kit includes a manufacturer’s one-year money-back guarantee and seven-year replacement warranty, one interface valve, one 3 l (0.8 gal.) reservoir bag, 1.8 m (6') of 6 mm (¼") I.D. suction hose, and 1.8 m (6') of 19 mm (¾") evacuation tubing.
Frequently Bought Together