Axygen® Mini Tube Systems, Corning
# de catalogue 89005-570
Fournisseur: Corning
Product available on GSA Advantage®
Spécifications
- Sterility:Non sterile
- Volume:0.65 ml
- Description:Single Tube
- Packaging:Bulk
- Supplier No.:MTS-06-C
Spécifications
A propos de cet article
PP, 100% virgin. The Axygen 96-well mini tube system is designed to work with 8-strip or 12-strip plug caps.
- Compatible with Beckman, Tecan, Zymark and other robotic workstations
- One-way lid design with etched in alphnumeric grid ensures positive sample identification
- Moulded in condensation rings on cover minimise risk of cross-contamination
Customers Who Bought This Also Bought