ContactUsLinkComponent
VWR® Reclosable UV-Protectant Amber Bags
Catalogue # 89005-330
Fournisseur:  VWR International
undefined
VWR® Reclosable UV-Protectant Amber Bags
Catalogue # 89005-330
Fournisseur:  VWR International
Spécifications
  • Cat. no.:
    89005-330
  • W×L:
    203×355 mm (8×14")
Spécifications
À propos de cet article
Ideal for storage and transport of light-sensitive products or medications.
  • Re-closable design
  • Amber tint protects against UV radiation
  • Easy to identify with amber tint
Re-closable 3 mil bags feature a wide, white zipper to allow for repeated use and provides protection against dust and moisture. Amber tint makes bags easily identifiable.
Certifications: Bags meet light transmission rates between the ranges of full shade at DIN 53775 and DIN 53387, as well as reduced shade at DIN 53375.
Frequently Bought Together