ContactUsLinkComponent
CLOSURE S/T UREA 24-400 PK144
Screw Caps with Rubber Liners, Kimble Chase, DWK Life Sciences
# de catalogue 89003-436
CLOSURE S/T UREA 24-400 PK144
Screw Caps with Rubber Liners, Kimble Chase, DWK Life Sciences
# de catalogue 89003-436
Spécifications
  • Cap Size:
    24-400
  • Color:
    White
  • Description:
    Urea Screw Thread Caps
  • Cat. no.:
    89003-436
  • Liner Material:
    PTFE-Faced Foam-Backed Rubber
  • Supplier No.:
    73802U-24400
Spécifications
A propos de cet article
Urea screw thread caps with PTFE-faced foam-backed rubber liners
Ideal for use with scintillation vials.
Customers Who Bought This Also Bought