Screw Caps with Rubber Liners, Kimble Chase, DWK Life Sciences
# de catalogue 89003-436
Fournisseur: DWK Life Sciences (KIMBLE)
Spécifications
- Cap Size:24-400
- Color:White
- Description:Urea Screw Thread Caps
- Cat. no.:89003-436
- Liner Material:PTFE-Faced Foam-Backed Rubber
- Supplier No.:73802U-24400
Spécifications
A propos de cet article
Urea screw thread caps with PTFE-faced foam-backed rubber liners
Ideal for use with scintillation vials.
Customers Who Bought This Also Bought