ContactUsLinkComponent
KIMAX* Brand Separatory Funnel, Squibb, Pear-Shape, PTFE Stopcock, Ground Glass Stopper
KIMAX® Separatory Funnels, Squibb, Pear-Shaped, Kimble Chase, DWK Life Sciences
Catalog # 89002-106
undefined
KIMAX® Separatory Funnels, Squibb, Pear-Shaped, Kimble Chase, DWK Life Sciences
Catalog # 89002-106
Specifications
  • Capacity:
    500 ml
  • Description:
    With [ST] ground glass stopper and PTFE stopcock
  • Cat. no.:
    89002-106
  • ST Stopper No.:
    27
  • ST Stopcock Size:
    4
  • Supplier No.:
    29048F-500
Specifications
About this item
Funnels are designed with a large lower stem diameter to allow more complete separation.
  • Pear-shaped
  • Autoclavable
Constructed with heavy walls, a fire-polished and chip-resistant stem, and heavily beaded rim.
Certifications: Designed from ASTM Specification E1096, Type 4 requirements. Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Delivery: Supplied with a Standard Taper ground glass stopper and an autoclavable PTFE stopcock plug.
Frequently Bought Together