KIMAX® Separatory Funnels, Squibb, Pear-Shaped, Kimble Chase, DWK Life Sciences
Catalog # 89002-106
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Specifications
- Capacity:500 ml
- Description:With [ST] ground glass stopper and PTFE stopcock
- Cat. no.:89002-106
- ST Stopper No.:27
- ST Stopcock Size:4
- Supplier No.:29048F-500
Specifications
About this item
Funnels are designed with a large lower stem diameter to allow more complete separation.
- Pear-shaped
- Autoclavable
Constructed with heavy walls, a fire-polished and chip-resistant stem, and heavily beaded rim.
Certifications: Designed from ASTM Specification E1096, Type 4 requirements. Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Delivery: Supplied with a Standard Taper ground glass stopper and an autoclavable PTFE stopcock plug.
Frequently Bought Together