Order Entry
ContactUsLinkComponent
KIMAX* Culture Tube with Rubber-Lined Screw Cap
KIMAX® Culture Tubes, Reusable, with Screw Cap, Kimble Chase
# de catalogue 89001-464
KIMAX* Culture Tube with Rubber-Lined Screw Cap
KIMAX® Culture Tubes, Reusable, with Screw Cap, Kimble Chase
# de catalogue 89001-464

Spécifications

  • Cap Size:
    15-415
  • Capacity:
    12 ml
  • Ø:
    16 mm
  • Height:
    100 mm
  • Supplier No.:
    45066 16100

Spécifications

A propos de cet article

Tubes with uniform wall thickness for maximum heat transfer and chemical resistance.

Tubes feature a marking spot and a threaded phenolic screw cap that can be loosened for autoclaving. Tubes are available with rubber-lined caps or PTFE-faced rubber-lined caps.

Certifications: Tubes with rubber-lined caps are designed from ASTM Specification E982, Type VI, Class A requirements. Tubes with PTFE-faced rubber-lined caps are designed from ASTM Specification E982, Type VI, Class B requirements.
Ordering information: Cap is supplied unattached.
Customers Who Bought This Also Bought