Order Entry
ContactUsLinkComponent
KIMAX* Test Tube
KIMAX® Test Tubes, Reusable, Borosilicate Glass, with Glass Stopper, Kimble Chase
# de catalogue 89000-866
KIMAX* Test Tube
KIMAX® Test Tubes, Reusable, Borosilicate Glass, with Glass Stopper, Kimble Chase
# de catalogue 89000-866

Spécifications

  • Capacity:
    26 ml
  • Ø:
    19 mm
  • Height:
    150 mm
  • ST Stopper No.:
    13
  • Supplier No.:
    45100-19150

Spécifications

A propos de cet article

Test tubes made from KG-33 borosilicate glass with a [ST] top and a [ST] glass stopper.

Uniform wall thickness for maximum service. Tubes incorporate a durable white ceramic marking spot. Designed from ASTM Specification E982, Type III requirements.

Customers Who Bought This Also Bought