Order Entry
ContactUsLinkComponent
FACESHIELD CLEAR UV PROTECT
Polycarbonate Face Shields, Oberon
Catalog # 87001-674
Supplier:  Oberon
FACESHIELD CLEAR UV PROTECT
Polycarbonate Face Shields, Oberon
Catalog # 87001-674
Supplier:  Oberon

Specifications

  • Description:
    Face Shield with Ratchet Head Gear
  • Cat. no.:
    87001-674
  • Supplier No.:
    179R

Specifications

About this item

The clear UV Protective Series face shields provide 100% protection from UV energy from 200 to 400nm.

  • Scratch resitant outside
  • Antifog inside

Not suitable for electrical arc flash protection, not intended for high intensity laser type applications.

Certifications: Meets ANSI Z87.1-2010 standards.
Customers Who Bought This Also Bought