ContactUsLinkComponent
VWR* 1.2mL Sample Library Tubes
Accessories for VWR® 1.2 ml Sample Library Tubes and Closures
# de catalogue 83009-680
Fournisseur:  VWR International
   Sustainable Packaging
   Low Manufacturing Impact
VWR* 1.2mL Sample Library Tubes
Accessories for VWR® 1.2 ml Sample Library Tubes and Closures
# de catalogue 83009-680
Fournisseur:  VWR International
   Sustainable Packaging
   Low Manufacturing Impact
Spécifications
  • Description:
    8-Well Tube Strips
  • Environmentally Preferable:
    Sustainable Packaging,
    Low Manufacturing Impact
  • Cat. no.:
    83009-680