VITLAB® Laboratory Bottles, LDPE, Narrow Mouth, with Screw Caps, PP, BrandTech
# de catalogue 83009-071
Fournisseur: Brandtech
Spécifications
- Cap Size:GL25
- Capacity:250 ml (8.5 oz.)
- Dimensions:58 Dia. x 135 H, mm
- Cat. no.:83009-071
- Supplier No.:V94989
Spécifications
A propos de cet article
Round-shouldered general-use low-density polyethylene laboratory bottles come with polypropylene screw caps.
- LDPE with PP screw caps
- Round shoulder design
- Narrow mouth
Bottles provide excellent chemical resistance to aliphatic alcohols, aldehydes, alkalis, and acids.
Ordering information: For replacement caps, see 83009-032 series.
Customers Who Bought This Also Bought