ContactUsLinkComponent
Polypropylene Graduated Pitcher
VITLAB® Graduated Pitchers, Polypropylene, BrandTech
# de catalogue 83008-742
Fournisseur:  Brandtech
Polypropylene Graduated Pitcher
VITLAB® Graduated Pitchers, Polypropylene, BrandTech
# de catalogue 83008-742
Fournisseur:  Brandtech
Spécifications
  • Capacity:
    1 L (34 oz.)
  • Graduation Interval:
    20 ml
  • Cat. no.:
    83008-742
  • Supplier No.:
    V442081
Spécifications
A propos de cet article
Made of durable, translucent polypropylene.
  • Pitchers with molded graduations areavailable with or without overprint
  • Nesting pitchers have screened overprint
  • Blue printing provides high contrast with many liquids
Standard graduated pitchers have molded graduations with or without a blue overprint for high legibility. Nesting pitchers conserve bench and storage space and have blue screened graduations for easy, accurate measurements.
Customers Who Bought This Also Bought