Specifications
- Format:MICROLON™ 600
- Sterility:Sterile
- Color:Clear
- Cat. no.:82051-576
- Surface Treatment:High binding
- Bottom Color:Clear
- Supplier No.:675061
Specifications
About this item
Polystyrene half area microplates are compatible with manual and automated pipetting, and dispensing and reading devices used with standard 96-well plates.
- Polystyrene construction
- Chimney-well design helps reduce cross-contamination
- Clear film bottoms
- Suitable for cell-based assays
This design allows up to a 50% sample and reagent reduction and the flat bottoms allow stackability with or without lids
Microplates with clear bottoms feature micro-thin µClear® bottoms for exceptional optical properties and standardized path-length, eliminating the need for path-length determinations. MICROLON™ formats are ideal for consistent performance in binding assays. White LUMITRAC™ plates are ideal for luminescence assays, while black FLUOTRAC™ plates are suitable for fluorescence assays. Both formats are also suitable for cell-based assays.
The chimney style well design reduces the risk of cross-contamination and aids sealing by cap mat, thermal sealer, or adhesive sealer.