ContactUsLinkComponent
HALF-AREA
96-Well Half Area Microplates, Greiner Bio-One
Catalog # 82051-576
Supplier:  Greiner Bio-One
HALF-AREA
96-Well Half Area Microplates, Greiner Bio-One
Catalog # 82051-576
Supplier:  Greiner Bio-One
Specifications
  • Format:
    MICROLON™ 600
  • Sterility:
    Sterile
  • Color:
    Clear
  • Cat. no.:
    82051-576
  • Surface Treatment:
    High binding
  • Bottom Color:
    Clear
  • Supplier No.:
    675061
Specifications
About this item
Polystyrene half area microplates are compatible with manual and automated pipetting, and dispensing and reading devices used with standard 96-well plates.
  • Polystyrene construction
  • Chimney-well design helps reduce cross-contamination
  • Clear film bottoms
  • Suitable for cell-based assays
This design allows up to a 50% sample and reagent reduction and the flat bottoms allow stackability with or without lids
Microplates with clear bottoms feature micro-thin µClear® bottoms for exceptional optical properties and standardized path-length, eliminating the need for path-length determinations. MICROLON™ formats are ideal for consistent performance in binding assays. White LUMITRAC™ plates are ideal for luminescence assays, while black FLUOTRAC™ plates are suitable for fluorescence assays. Both formats are also suitable for cell-based assays.
The chimney style well design reduces the risk of cross-contamination and aids sealing by cap mat, thermal sealer, or adhesive sealer.
Customers Who Bought This Also Bought