Inoculating Loops and Needles, Sterile, Greiner Bio-One
# de catalogue 82051-144
Fournisseur: Greiner Bio-One
Spécifications
- Length:20 cm (77/8")
- Color:Blue
- Description:10 µL Inoculating Loops
- Cat. no.:82051-144
- Packaging:Individually Packed
- Supplier No.:731171
Spécifications
A propos de cet article
Inoculating loops allow easier bacterial collection and inoculation.
- Color-coded
- Polystyrene construction
- Sterile
Inoculating needles are ideal for picking up single colonies.
Customers Who Bought This Also Bought