ContactUsLinkComponent
96-Well Plate
96-Well Black and White Microplates, Polystyrene, Greiner Bio-One
# de catalogue 82050-750
Fournisseur:  Greiner Bio-One
Product available on GSA Advantage®
96-Well Plate
96-Well Black and White Microplates, Polystyrene, Greiner Bio-One
# de catalogue 82050-750
Fournisseur:  Greiner Bio-One
Product available on GSA Advantage®
Spécifications
  • Sterility:
    Sterile
  • Color:
    White
  • Surface Treatment:
    High binding
  • Working Well Volume:
    382 µl
  • Maximum Well Volume:
    25–340 µL
  • Supplier No.:
    655094
Spécifications
A propos de cet article
Black and white microplates are manufactured from virgin polystyrene resin and are designed to meet industry standard dimension specifications to facilitate compatibility with microplate handling and robotics.
  • Free of RNase, DNase, and human DNA
  • Nonpyrogenic
  • Virgin polystyrene construction
  • Solid bottoms or clear bottoms
All of the black and white plates have square chimney wells with flat bottoms
LUMITRAC™ and FLUOTRAC™ formats are offered to allow a wide variety of applications, including cell-based assays. White LUMITRAC™ plates are for luminescence assays and black FLUOTRAC™ plates are for fluorescence assays.
Plates are available with solid or clear bottoms. Clear bottom plates are manufactured with µClear™ micro-thin polystyrene film, and are suggested for assays designed for luminescence and fluorescence detection in which imaging from the bottom is required. Plates are supplied without lids.
These 96-well polystyrene microplates plates are free of detectable RNase, DNase, human DNA, and pyrogens. All plates have a footprint conforming to the ANSI/SBS 1-2004 Standard.
Customers Who Bought This Also Bought