VWR® Microscope Slides and Plastic Coverslips
# de catalogue 82027-788
Fournisseur: VWR International
Small Business Enterprise
Spécifications
- Description:Slide
- L×W:75×25 mm (3×1")
- Thickness:0.45 - 0.5 mm
- Cat. no.:82027-788
Spécifications
A propos de cet article
Efficient, disposable slides are manufactured from optically clear vinyl.
- Made of clear vinyl plastic
- Refractive index as glass
- Unbreakable and safer than glass
- Used in biological and medical labs, as well as educational applications
Efficient, disposable slides are manufactured from optically clear vinyl.
Slides will not break, scratch, or cut fingers. Ideal for wet mounts. Micro coverslips are unbreakable and non-curling.
Customers Who Bought This Also Bought