ContactUsLinkComponent
VWR 9.25 inch PIPET BOX
VWR® Pipette Sterilization Boxes, Stainless Steel
Catalog # 82027-604
Supplier:  VWR International
VWR 9.25 inch PIPET BOX
VWR® Pipette Sterilization Boxes, Stainless Steel
Catalog # 82027-604
Supplier:  VWR International
Specifications
  • Description:
    Square (Pasteur)
  • Dimensions:
    5.1W×5.1D×23.5H cm (2×2×9¼")
  • Cat. no.:
    82027-604
Specifications
About this item
Pipette sterilization boxes are designed to secure and protect pipettes during sterilization processes.
  • Stainless steel construction
  • High corrosion resistance
  • Compatible with a variety of pipette types
Stainless steel construction ensures high corrosion resistance. Multiple box styles accommodate a variety of pipette types.
Customers Who Bought This Also Bought