ContactUsLinkComponent
VWR* Dissecting Scissors, 5 inch
VWR® Dissecting Scissors, Curved Tip, 5"
# de catalogue 82027-586
Fournisseur:  VWR International
VWR* Dissecting Scissors, 5 inch
VWR® Dissecting Scissors, Curved Tip, 5"
# de catalogue 82027-586
Fournisseur:  VWR International
Spécifications
  • Cat. no.:
    82027-586
  • Length:
    127 mm (5")
Spécifications
A propos de cet article
VWR® Dissecting Scissors are stainless steel with fine, bent tips.
  • Stainless steel fabrication
  • Curved tips
Dissecting scissors are used for a variety of applications and are available in different lengths.
Customers Who Bought This Also Bought