Order Entry
ContactUsLinkComponent
VWR CRUCIBLE NICKEL 50ML
VWR® Nickel Crucibles and Covers
Catalog # 82027-560
Supplier:  VWR International
VWR CRUCIBLE NICKEL 50ML
VWR® Nickel Crucibles and Covers
Catalog # 82027-560
Supplier:  VWR International

Specifications

  • Capacity:
    50 ml
  • Cat. no.:
    82027-560
  • Diameter ×H:
    46.5 x 49.5 mm

Specifications

About this item

High form crucibles and covers are drawn from one-piece, pure sheet nickel.

They are highly resistant to dilute alkalies, and can withstand temperatures up to 800°C (1472°F).

Ordering information: Crucibles and covers are sold separately.
Customers Who Bought This Also Bought