VWR® Round/Flat Spatulas, Stainless Steel
Catalog # 82027-528
Supplier: VWR International
Specifications
- Length:17.8 cm (7")
- Cat. no.:82027-528
- Blade Length:—
Specifications
About this item
Ideal for transferring, collecting, mixing, or handling chemical or biological samples.
- Rustproof
- Nonsticking
- Multipurpose design
The versatile design of these stainless steel models enables use as a spatula, spoon, scraper, or stirring rod. Spatulas feature one rounded end and one flat end. These multi-purpose items are rustproof and feature non stick surfaces. Spatula 82027-494 features heavy-duty construction.
Customers Who Bought This Also Bought