VWR® Micro Spoon, Stainless Steel
Catalog # 82027-526
Supplier: VWR International
Specifications
- Length:16.5 cm (6½")
Specifications
About this item
Designed for use in the transfer and handling of small chemical or biological samples.
- Rustproof
- Stainless steel construction
The rustproof micro spoon is constructed of stainless steel and features a plastic handle. Substances will not stick to spoon surface.
Frequently Bought Together