VWR® Hayman Style Spatulas, Stainless Steel
# de catalogue 82027-522
Fournisseur: VWR International
Spécifications
- Length:16.5 cm (6¹/₂")
- Description:Rounded spatula/spoon
Spécifications
A propos de cet article
Stainless steel spatulas ideal for applications involving the transfer or mixing of small sample amounts.
- Efficient design
- Stainless steel construction
Model 82027-522 features a round spatula on one end and a spoon on the other. Model 82027-524 features a tapered, bent-tip spatula on one end and a spoon on the other. Efficient design reduces the risk of sample loss.
Customers Who Bought This Also Bought