VWR® Round/Tapered Micro Spatulas, Stainless Steel
# de catalogue 82027-518
Fournisseur: VWR International
Spécifications
- Length:20.3 cm (8")
- Cat. no.:82027-518
- Blade Length:203 mm (8")
Spécifications
A propos de cet article
Ideal for small quantity applications such as sample transfer into test tubes or microcentrifuge tubes.
- Mirror finish
- Rustproof
- Non-sticking
Stainless steel spatulas feature one rounded end and one tapered end, allowing for a variety of transfer, mixing, or handling applications.
Customers Who Bought This Also Bought