ContactUsLinkComponent
VWR* PTFE-Coated Forceps
VWR® PTFE-Coated Forceps
# de catalogue 82027-454
Fournisseur:  VWR International
VWR* PTFE-Coated Forceps
VWR® PTFE-Coated Forceps
# de catalogue 82027-454
Fournisseur:  VWR International
Spécifications
  • Length:
    114 mm (41/2")
  • Cat. no.:
    82027-454
Spécifications
A propos de cet article
Stainless steel forceps are PTFE-coated.
  • Stainless steel construction
  • Chemically resistant
  • Nonsticking and non-corroding
Forceps are coated with PTFE to imbue forceps with heat and chemical resistance.
Customers Who Bought This Also Bought