ContactUsLinkComponent
VWR* Tissue Forceps
VWR® Tissue Forceps
# de catalogue 82027-446
Fournisseur:  VWR International
VWR* Tissue Forceps
VWR® Tissue Forceps
# de catalogue 82027-446
Fournisseur:  VWR International
Spécifications
  • Length:
    152 mm (6")
  • Cat. no.:
    82027-446
Spécifications
A propos de cet article
Tissue forceps are specially designed to grasp tissue with minimal damage to the organic material.
  • Stainless steel construction
  • Straight tips
  • Easy to use
These tools are available in a variety of designs, lengths, and specific uses for laboratory procedures.
Customers Who Bought This Also Bought