ContactUsLinkComponent
VWR* Gooseneck Forceps
VWR® Gooseneck Forceps
Catalog # 82027-444
Supplier:  VWR International
VWR* Gooseneck Forceps
VWR® Gooseneck Forceps
Catalog # 82027-444
Supplier:  VWR International
Specifications
  • Length:
    152 mm (6")
  • Cat. no.:
    82027-444
Specifications
About this item
These stainless steel forceps are used for dissecting, handling insects, or positioning specimens for pinning.
  • Stainless steel construction
  • Serrated tips
The forceps have serrated tips and a handle with a gooseneck shank.
Customers Who Bought This Also Bought