VWR® Gooseneck Forceps
Catalog # 82027-444
Supplier: VWR International
Specifications
- Length:152 mm (6")
- Cat. no.:82027-444
Specifications
About this item
These stainless steel forceps are used for dissecting, handling insects, or positioning specimens for pinning.
- Stainless steel construction
- Serrated tips
The forceps have serrated tips and a handle with a gooseneck shank.
Customers Who Bought This Also Bought