VWR® Micro-Adson Forceps
# de catalogue 82027-430
Fournisseur: VWR International
Spécifications
- Length:127 mm (5")
- Cat. no.:82027-430
Spécifications
A propos de cet article
Micro-Adson forceps are for holding and manipulating delicate tissues.
- Stainless steel construction
- Micro-Adson type
- Serrated jaws
These forceps have a wide, flat thumb grasp area that is commonly serrated. The jaws are short and the tips are extremely narrow.
Customers Who Bought This Also Bought