ContactUsLinkComponent
VWR* Micro-Adson Forceps
VWR® Micro-Adson Forceps
# de catalogue 82027-430
Fournisseur:  VWR International
VWR* Micro-Adson Forceps
VWR® Micro-Adson Forceps
# de catalogue 82027-430
Fournisseur:  VWR International
Spécifications
  • Length:
    127 mm (5")
  • Cat. no.:
    82027-430
Spécifications
A propos de cet article
Micro-Adson forceps are for holding and manipulating delicate tissues.
  • Stainless steel construction
  • Micro-Adson type
  • Serrated jaws
These forceps have a wide, flat thumb grasp area that is commonly serrated. The jaws are short and the tips are extremely narrow.
Customers Who Bought This Also Bought