VWR® Rochester-Pean Hemostatic Forceps
# de catalogue 82027-418
Fournisseur: VWR International
Spécifications
- Length:159 mm (61/4")
- Description:Straight
- Cat. no.:82027-418
Spécifications
A propos de cet article
Rochester-Pean Hemostatic Forceps are used for clamping larger tissue and vessels for hemostasis.
- Stainless steel construction
- Reusable
- Non-sterile
- Premium grade
Horizontal serrations help ensure forceps remain clamped tight.
Customers Who Bought This Also Bought