VWR® Hemostatic Forceps
Catalog # 82027-414
Supplier: VWR International
Specifications
- Description:Straight
- Length:127 mm (5")
- Cat. no.:82027-414
Specifications
About this item
Hemostatic forceps are used in many surgical procedures, usually to control bleeding or the flow of liquids in tubing.
- Stainless steel fabrication
These forceps have ring handles just like scissors, and ratcheted handles can be locked in multiple positions in order to maintain variable levels of constant pressure.
Customers Who Bought This Also Bought