ContactUsLinkComponent
VWR* Dissecting Forceps
VWR® Dissecting Forceps, Fine Tip, Serrated
# de catalogue 82027-408
Fournisseur:  VWR International
VWR* Dissecting Forceps
VWR® Dissecting Forceps, Fine Tip, Serrated
# de catalogue 82027-408
Fournisseur:  VWR International
Spécifications
  • Length:
    114 mm (41/2")
  • Description:
    Straight
  • Cat. no.:
    82027-408
Spécifications
A propos de cet article
VWR® Dissecting Forceps are stainless steel with fine, serrated tips.
  • Stainless steel fabrication
  • Serrated, fine tips
Dissecting forceps are used for a variety of applications and are available in different lengths.
Customers Who Bought This Also Bought