VWR® Dissecting Forceps, Superfine Tip
# de catalogue 82027-404
Fournisseur: VWR International
Spécifications
- Length:108 mm (41/4")
- Description:Curved
- Cat. no.:82027-404
Spécifications
A propos de cet article
VWR® Dissecting Forceps are stainless steel with superfine tips.
- Stainless steel fabrication
- Superfine tips
Dissecting forceps are used for a variety of applications and are available in different lengths.
Customers Who Bought This Also Bought