VWR® Tongs, PTFE-Coated
# de catalogue 82027-376
Fournisseur: VWR International
Spécifications
- Length:22.9 cm (9")
- Cat. no.:82027-376
Spécifications
A propos de cet article
Designed to handle hot, cold, corrosive, or sticky materials without scratching or damaging container exteriors.
- Stainless steel construction
- PTFE coating
- Nonsticking and non-corrosive
Contoured jaw enables the handling of crucibles and other objects. Tongs are used for handling chemically active materials.
Customers Who Bought This Also Bought