ContactUsLinkComponent
VWR PTFE COSTED TONGS
VWR® Tongs, PTFE-Coated
# de catalogue 82027-376
Fournisseur:  VWR International
VWR PTFE COSTED TONGS
VWR® Tongs, PTFE-Coated
# de catalogue 82027-376
Fournisseur:  VWR International
Spécifications
  • Length:
    22.9 cm (9")
  • Cat. no.:
    82027-376
Spécifications
A propos de cet article
Designed to handle hot, cold, corrosive, or sticky materials without scratching or damaging container exteriors.
  • Stainless steel construction
  • PTFE coating
  • Nonsticking and non-corrosive
Contoured jaw enables the handling of crucibles and other objects. Tongs are used for handling chemically active materials.
Customers Who Bought This Also Bought