ContactUsLinkComponent
VWR* Crucible Tongs
VWR® Crucible Tongs
# de catalogue 82027-368
Fournisseur:  VWR International
VWR* Crucible Tongs
VWR® Crucible Tongs
# de catalogue 82027-368
Fournisseur:  VWR International
Spécifications
  • Length:
    45.7 cm (18")
  • Cat. no.:
    82027-368
Spécifications
A propos de cet article
Crucible tongs are designed for use in handling a wide variety of laboratory apparatus.
  • Nickel-plated stainless steel fabrication
The 24.1cm model is a general-purpose tong with serrated tips. The 45.7cm model features nickel-plated steel construction and an extra-long design.
Customers Who Bought This Also Bought