VWR® 60-Minute Mechanical Timer
# de catalogue 82023-840
Fournisseur: VWR International
Spécifications
- Cat. no.:82023-840
- W×H:57×57 mm (2.2×2.2")
- Accuracy:3 minutes
Spécifications
A propos de cet article
Dial timer has a timing capacity of 60 minutes.
- Counts down from 60 minutes
- Audible alarm
- Highly visible red arrow
Customers Who Bought This Also Bought