VWR® Micro Slide Trays, Cardboard
# de catalogue 82020-913
Fournisseur: VWR International
Spécifications
- Cat. no.:82020-913
- No. of Places:20
- L×W×H:20.8 x 34.1 x 9 cm (83/16 x 137/16 x 31/2")
Spécifications
A propos de cet article
VWR® Micro Slide Trays accommodate 20 or 30 standard 25x75 mm (1x3") slides.
- Sturdy cardboard construction
- Recessed compartments protect specimens
- Designed to allow easy slide insertion and removal
The horizontal compartments are recessed, protecting the specimens when the cover is closed. Compartments feature a thumb cut for easy slide removal.
Customers Who Bought This Also Bought