ContactUsLinkComponent
VWR SLIDE MAILR CARDBRD 1 PK25
VWR® Disposable Slide Mailers
# de catalogue 82020-908
Fournisseur:  VWR International
VWR SLIDE MAILR CARDBRD 1 PK25
VWR® Disposable Slide Mailers
# de catalogue 82020-908
Fournisseur:  VWR International
Spécifications
  • Description:
    One-Place Slide Mailer
  • W×D×H:
    40 x 5 x 95 mm (19/16 x 3/16 x 311/16")
Spécifications
A propos de cet article
Heavy cardboard holders accommodate one or two standard 25x75 mm (1x3") slides.
  • Cardboard construction
  • Slides are easy to remove
Thumb grooves allow for easy removal of slides.
Customers Who Bought This Also Bought