Spécifications
- W×L:25.4 x 33 cm (10 x 13")
- Cat. no.:82020-458
Spécifications
A propos de cet article
These all-purpose wipes are versatile cloth-like wipes that are safe for skin contact, with no rinse required.
- Soft cloth-like material
- No rinse, skin formula
- Alcohol-free
A super absorbent layer of air-lay cellulose provides both softness and strength. Wipes are sensitive and soft enough for use on skin, and they are versatile enough for use on surfaces.