ContactUsLinkComponent
VWR* Disposable Washcloths; Disposable Wipes; Disposable Cloths; Wipes; Cleansing Wipes; Cleansing Cloths; Cleansing Cloths; Cellulose Cloths; Cloths; Wet Wipes; Incontinence Wipes; Absorbent Wipes
VWR® All-Purpose Disposable Cloth-Like Wipes
# de catalogue 82020-458
Fournisseur:  VWR International
VWR* Disposable Washcloths; Disposable Wipes; Disposable Cloths; Wipes; Cleansing Wipes; Cleansing Cloths; Cleansing Cloths; Cellulose Cloths; Cloths; Wet Wipes; Incontinence Wipes; Absorbent Wipes
VWR® All-Purpose Disposable Cloth-Like Wipes
# de catalogue 82020-458
Fournisseur:  VWR International
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • W×L:
    25.4 x 33 cm (10 x 13")
  • Cat. no.:
    82020-458
Spécifications
A propos de cet article
These all-purpose wipes are versatile cloth-like wipes that are safe for skin contact, with no rinse required.
  • Soft cloth-like material
  • No rinse, skin formula
  • Alcohol-free
A super absorbent layer of air-lay cellulose provides both softness and strength. Wipes are sensitive and soft enough for use on skin, and they are versatile enough for use on surfaces.
Customers Who Bought This Also Bought