VWR® Enviro-Safe® Environmentally Friendly Liquid-In-Glass Pocket Thermometers
# de catalogue 82007-990
Fournisseur: VWR International
Spécifications
- Accuracy:±1 °C
- Length:160 mm (65/16")
- Description:Enviro Safe Pocket Thermometer
- Cat. no.:82007-990
- Packaging:Closed Plastic Case
- Divisions:1 °C
- Fill Solution:Enviro-Safe® liquid
- Range:−10...+110 °C
- Immersion:Total
Spécifications
A propos de cet article
EnviroKleen™ certified thermometers feature green, non-toxic, biodegradable, Enviro-Safe® liquid against lead-free, white back glass.
Environmentally safe thermometers are portable enough to go anywhere and can be used for most applications.
- Total immersion pocket thermometers are a compact 160mm (6.3”) long
- Comes with a case to protect from breakage
- Ideal for field studies, waste water treatment and any “on-the-go” applications
- Manufactured and tested by our USA quality team
Filled with biodegradable, non-hazardous liquid and dye, Enviro-Safe thermometers increase lab safety with no mercury problems to worry about. Compact and durable. The white background and green liquid color make them easy to read.
All Enviro-Safe thermometers have permanently fused markings. A statement of accuracy is provided to indicate instrument traceability to standards provided by the National Institute of Standards and Technology (NIST) and Deutscher Kalibrierdienst (DKD).
Certifications: Traceable to NIST.
Ordering information: Includes a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability.
Customers Who Bought This Also Bought