VWR® Microscope Slide Boxes for 25 Slides
# de catalogue 82003-426
Fournisseur: VWR International
Spécifications
- Color:White
- Cat. no.:82003-426
Spécifications
A propos de cet article
These heavy-duty, two-piece slide boxes feature cork lining and store up to 25 standard 25x75 mm (1x3") slides.
- Heavy-duty design
- Color-coded for easy identification
Boxes stack securely on top of each other. Ideal for storage or transport. Dimensions: 14Lx8.9Wx3.2Dcm (51/2x31/2x11/4").
Customers Who Bought This Also Bought