Specifications
- Type:Metal Luer Lock Hub
- Volume:50 ml
- Description:Glass Syringes
- Cat. no.:80089-576
- For Use With:Laboratory and Veterinary Use Only
- Packaging:1/box
- Supplier No.:GL50
Specifications
About this item
Fortuna Optima syringes are specialty syringes made from very fine borosilicate glass.
- Borosilicate glass
- Interchangeable pistons and barrels
- Available with Luer Slip Tip or Luer Lock Tip
These syringes are manufactured in Germany from very fine borosilicate glass with interchangeable pistons and barrels. They feature amber graduations and have Luer lock and slip tips.