Axygen® Pipette Tip 200 µl with Graduation Marks, Corning
# de catalogue 80086-098
Fournisseur: Corning
Product available on GSA Advantage®
Spécifications
- Volume:200 µl
- Tip Color:Yellow
- Packed:Rack
- Tip Style:Universal
- Supplier No.:TE-204-Y-R
Spécifications
A propos de cet article
Axygen brand 200 µl tips are designed to fit Eppendorf-Style single-channel and multi-channel pipettors.
Flexible walls and a series of internal sealing rings ensure a secure fit and less force required to load and eject. Accuracy and precision testing is performed for every manufacturing lot using the finest pipettors in the industry. These tips have graduation marks at 10, 50 and 100 µl for your accurate pipetting.
Customers Who Bought This Also Bought