ContactUsLinkComponent
RACK PCR TUBE SPECTRUM CS25
Axygen® PCR Tube Storage Racks, Corning
Catalog # 80086-074
Supplier:  Corning
Product available on GSA Advantage®
   Low Manufacturing Impact
undefined
Axygen® PCR Tube Storage Racks, Corning
Catalog # 80086-074
Supplier:  Corning
Product available on GSA Advantage®
   Low Manufacturing Impact
Specifications
  • Color:
    Spectrum Pack
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    I-RECs, ISO 14001 - Reynosa
  • Cat. no.:
    80086-074
  • Supplier No.:
    R-96-PCR-FSP
Specifications
About this item
PP with lid.
  • Fluorescent color base with translucent lid
  • Durable, chemically inert, and can withstand extended duration at sub-zero temperatures
Designed to hold 0.2, and 0.5 ml PCR tubes.
Ordering information: Assorted pack includes one each of blue, green, violet, red and yellow.
Frequently Bought Together