Axygen® Pipette Filter Tip, Axygen®, Corning
# de catalogue 80086-062
Fournisseur: Corning
Product available on GSA Advantage®
Spécifications
- Volume:1200 µl
- Cat. no.:80086-062
- Sterile:+
- Supplier No.:TF-1200-C-R-S
Spécifications
A propos de cet article
Axygen brand filter tips allow free passage of air while preventing passage of aerosols.
The filters contain no leachable additives.
Customers Who Bought This Also Bought