VWR® Chlorobutyl Serum and Lyophilization Stoppers
# de catalogue 80079-870
Fournisseur: VWR International
Spécifications
- Size:13 mm
- Cat. no.:80079-870
Spécifications
A propos de cet article
Stoppers are molded from elastomeric formula 4432/50 Gray. This formula has extremely low levels of organic and metallic extractables and does not contain plasticizers, 2-Mercaptobenzothiazole, nitrosamine precursors, or natural rubber latex.
All stoppers are manufactured in cGMP facilities and tested for quality. Stoppers meet USP, Ph. Eur., and JP requirements.
Enhanced stoppers are also ready for sterilization without further preparation. They are packaged in double sterilizable bags designed for direct entry into sterilization units. Each shipment also includes a certificate of analysis.
Customers Who Bought This Also Bought