Axygen® ImpermaMat Sealing Mats, Corning
# de catalogue 80078-816
Fournisseur: Corning
Product available on GSA Advantage®
Spécifications
- Sterility:Non sterile
- Description:ImpermaMat™ sealing mat for 96-well deep well plates with square wells
- Material:Silicone
- Supplier No.:AM-2ML-SQ-IMP
Spécifications
A propos de cet article
Axygen® ImpermaMat sealing mats minimize evaporation in a wide range of temperatures in PCR and storage applications. ImpermaMats are chemically resistant, reusable sealing mats, that provide a high degree of confidence.
- Chemical resistance - can be used in any application requiring the presence of DMSO or similar solvent
- Contains no adhesives, no extractables to contaminate samples
- Tight seal to the plate, no sample evaporation or well-to-well contamination
- Usable within wide temperature range from –80 up to +121 °C
- High contrast alphanumeric grid for easy well identification
For storage applications involving more aggressive solvents, they contain no adhesives or extractable to contaminate samples. Made of research-grade silicone, they can be autoclaved, and following a bleach wash and ethanol rinse protocol, they can also be reused.
Customers Who Bought This Also Bought