ContactUsLinkComponent
Self-Standing Screw Cap Microtube with Cap
Axygen® Self-Standing Screw Top Microcentrifuge Tubes with Caps, Corning
Catalog # 80078-428
Supplier:  Corning
Product available on GSA Advantage®
Self-Standing Screw Cap Microtube  with Cap
Axygen® Self-Standing Screw Top Microcentrifuge Tubes with Caps, Corning
Catalog # 80078-428
Supplier:  Corning
Product available on GSA Advantage®
Specifications
  • Sterility:
    Sterile
  • Volume:
    2 ml
  • Description:
    Microtube
  • Cat. no.:
    80078-428
  • Maximum RCF:
    20000×g
  • Cap Color:
    Clear
  • Supplier No.:
    SCT-200-SS-C-S
Specifications
About this item
Microcentrifuge tubes serve a broad range of laboratory needs including collection, centrifugation, long-term storage, and OEM packaging and shipping of restriction enzymes or other DNA/RNA modifying reagents.
Threaded O-ring design ensures a uniform closure. Ribs on cap and tube are designed to fit the AxyRack™ microtube racks (see 10011-272 series) to allow single-handed opening and closing. MAXYMum Recovery™ tubes feature an ultra-smooth surface completely free of occlusions and cavities, virtually eliminating sample retention while improving accuracy. Certified RNase-, DNase-, and pyrogen-free.
Customers Who Bought This Also Bought